Order Entry
Sweden
ContactUsLinkComponent
 
Organic reference standards, betaxolos, Ph. Eur.
 :  EDQM
 
Organic reference standards, betaxolos, Ph. Eur.
 :  EDQM

 

  • CAS No.:
    63659-19-8
  • Formula:
    C₁₈H₃₀ClNO₃
  • Description:
    Betaxolol hydrochloride