Order Entry
Sweden
ContactUsLinkComponent
Penicillin : Streptomycin solution 100X
Penicillin : Streptomycin solution 100X
Catalog # 30-002-CI
Supplier:  Corning
CAS Number:  
undefined
Penicillin : Streptomycin solution 100X
Catalog # 30-002-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6
  • Seq:
    2455
  • Pk Info:
    6x 100 ml

Specifications

Specification Test Results

  • Penicillin G, potassium salt:
    10,000,000 U/L
  • Streptomycin sulfate:
    10.0 g/l

Specification Test Results

About this item

 :
 :GHS08
CAS Number:113-98-4 (Penicillin G potassium salt) - 3810-74-0 (Streptomycin sulfate)
 :3810-74-0