Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc sulphate heptahydrate
Zinc sulphate heptahydrate Ph. Eur., Macron Fine Chemicals™
Zinc sulphate heptahydrate Ph. Eur., Macron Fine Chemicals™
Zinc sulphate heptahydrate Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
Zinc sulphate heptahydrate Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:
7446-20-0
:
Formula:
ZnSO₄·7H₂O
MDL Number:
MFCD00149894
ADR:
9,III
:
287.54 g/mol
UN:
3077
Related Information
Documentation
Certificates
Search by Lot Number
Documentation