Specifications
- Antibody type:Primary
- Antigen name:Vestigial-like Family Member 2
- Antigen symbol:VGLL2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:245806
- Antigen synonyms:VITO1|VGL2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Vgl-2, also known as VITO-1, is a 317 amino acid protein that contains a domain through which it interacts with TEF-1, a protein that plays a role in controlling the expression of numerous genes. Specific to skeletal muscle, Vgl-2 is expressed highly in adult fast muscle and is expressed at lower levels in adult slow muscle and fetal skeletal muscle. During muscle differentiation, Vgl-2 mRNA levels increase and Vgl-2 translocates from the cytoplasm to the nucleus. Overexpression of Vgl-2 in MYOD-transfected 10T1/2 mouse embryonic fibroblasts increases expression of myosin heavy chain (MHC), which is a marker of terminal muscle differentiation. This evidence suggests that Vgl-2 is essential for muscle gene expression. There are two isoforms of Vgl-2 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: VGLL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat