Anti-OSTN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-9566R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Osteocrin
- Antigen symbol:OSTN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:344901
- Antigen synonyms:Osteocrin|MUSCLIN|OSTN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:83-133/133
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Appears to modulate osteoblastic differentiation. Could also function as an autocrine and paracrine factor linked to glucose metabolism in skeletal muscle (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OSTN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat