Anti-SRXN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8329R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Sulfiredoxin 1
- Antigen symbol:SRXN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:140809
- Antigen synonyms:Npn3|Sulfiredoxin 1 homolog (S. cerevisiae)|C20orf139|sulfiredoxin 1 homolog|dJ850E9.2|SRX1|YKL086W
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:76-98/137
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Contributes to oxidative stress resistance by reducing cysteine-sulfinic acid formed under exposure to oxidants in the peroxiredoxins PRDX1, PRDX2, PRDX3 and PRDX4. Does not act on PRDX5 or PRDX6. May catalyze the reduction in a multi-step process by acting both as a specific phosphotransferase and a thioltransferase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SRXN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat