Anti-COA3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-8116R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome c oxidase assembly factor 3
- Antigen symbol:COA3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:28958
- Antigen synonyms:MITRAC12|CCDC56|HSPC009
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:79-106/106
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Component of some MITRAC complex, a cytochrome c oxidase (COX) assembly intermediate complex that regulates COX assembly. MITRAC complexes regulate both translation of mitochondrial encoded components and assembly of nuclear-encoded components imported in mitochondrion. Required for efficient translation of MT-CO1 and mitochondrial respiratory chain complex IV assembly.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COA3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: