Order Entry
Sweden
ContactUsLinkComponent
Anti-HOXC9 Rabbit Polyclonal Antibody (Cy7®)
Anti-HOXC9 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-7982R-CY7
Leverantör:  Bioss
Anti-HOXC9 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-7982R-CY7
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Homeobox C9
  • Antigensymbol:
    HOXC9
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy7®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    3225
  • Antigen synonymer:
    HOX3B|HOX3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    150-200/260
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HOXC9
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat