Anti-BCL2L14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-7585R-A488
: Bioss
- Antibody type:Primary
- Antigen name:BCL2-Like 14
- Antigen symbol:BCL2L14
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:79370
- Antigen synonyms:Apoptosis facilitator Bcl-2-like protein 14|Apoptosis regulator Bcl-G|Bcl2-L-14|Bcl-G|BCLG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:280-292/327
- Purification:Purified by Protein A
- Pk:100 µl
Plays a role in apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BCL2L14
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat