Anti-IFNA10 Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-7024R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:Interferon, Alpha 10
- Antigen symbol:IFNA10
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:3446
- Antigen synonyms:IFN-alphaC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IFNA10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human