Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6338R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein C4
- Antigen symbol:APOC4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:346
- Antigen synonyms:Apolipoprotein C IV|APOC 4|ApoC IV|APOC4_HUMAN.|APO C4|APOC4|Apo CIV
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a lipid-binding protein belonging to the apolipoprotein gene family. The protein is thought to play a role in lipid metabolism. Polymorphisms in this gene may influence circulating lipid levels and may be associated with coronary artery disease risk. This gene is present in a cluster with other related apolipoprotein genes on chromosome 19. Naturally occurring read-through transcription exists between this gene and the neighboring downstream apolipoprotein C-II (APOC2) gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOC4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human