Anti-ID3 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-6229R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Inhibitor Of Differentiation 3
- Antigen symbol:ID3
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3399
- Antigen synonyms:bHLHb25|inhibitor of DNA binding 3 dominant negative helix-loop-helix Protein|HEIR-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
ID (inhibitor of DNA binding) HLH proteins lack a basic DNA-binding domain but are able to form heterodimers with other HLH proteins, thereby inhibiting DNA binding. ID-3 inhibits the binding of E2A-containing protein complexes to muscle creatine kinase E-box enhancer. May inhibit other transcription factors.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ID3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat