Order Entry
Sweden
ContactUsLinkComponent
 
Anti-AdV 5 E1A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-6136R-A488
 :  Bioss
Anti-AdV 5 E1A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-6136R-A488
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Adenovirus 5 E1A
  • Antigensymbol:
    AdV 5 E1A
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 488
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Virus
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Antigen synonymer:
    Protein BS69|Adenovirus 5 E1A-binding protein|MRD30|Bone morphogenetic protein receptor-associated molecule 1|Zinc finger MYND domain-containing protein 11|BRAM1|BS69|ZMYND11
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    180-230/289
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

The early region (E1) of the adenovirus genome, responsible for transforming activity, is localized within the left most 11% of the viral genome and consists of two transcriptional units E1A and E1B. E1A is sufficient for partial transformation and immortalization of primary cells. E1A gene products are necessary for normal levels of transcription of the other early regions of the adenovirus genome during productive infection and are able to either activate or repress the transcription of specific cellular genes. E1A forms specific complexes with cellular proteins including p105 causing inhibition of the cell cycle inducing arresting function of p105.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AdV 5 E1A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus