Anti-CA10 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6024R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 10
- Antigen symbol:CA10
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:56934
- Antigen synonyms:CARPX|CA-RPX|HUCEP-15
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Carbonic anhydrase X belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. This protein is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CA10
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat