Order Entry
Sweden
ContactUsLinkComponent
Anti-AIM2 Rabbit Polyclonal Antibody (Cy7®)
Anti-AIM2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5986R-CY7
Supplier:  Bioss
Anti-AIM2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5986R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    absent in melanoma 2
  • Antigen symbol:
    AIM2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    383619
  • Antigen synonyms:
    Ifi210,interferon-inducible protein AIM2|PYHIN4,CRYBG2,Gm1313
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    250-300/354
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Involved in innate immune response by recognizing cytosolic double-stranded DNA and inducing caspase-1-activating inflammasome formation in macrophages. Upon binding to DNA is thought to undergo oligomerization and to associate with PYCARD initiating the recruitment of caspase-1 precusrsor and processing of interleukin-1 beta and interleukin-18. Detects cytosolic dsDNA of viral and bacterial origin in a non-sequence-specific manner. Can also trigger PYCARD-dependent, caspase-1-independent cell death that involves caspase-8.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AIM2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat