Order Entry
Sweden
ContactUsLinkComponent
Anti-MATR3 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-MATR3 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5141R-CY5.5
Supplier:  Bioss
Anti-MATR3 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5141R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrin 3
  • Antigen symbol:
    MATR3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9782
  • Antigen synonyms:
    KIAA0723|MATR3_HUMAN|Matrin3|VCPDM.|MATR3|Matrin-3|MPD2|Matr3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Matrin 3 (MATR 3) is an internal nuclear matrix protein that may play a role in transcription or may interact with other nuclear matrix proteins to form the internal fibrogranular network. In association with the SFPQ-NONO heteromer MATR 3 may play a role in nuclear retention of defective RNAs. MATR3 forms part of complex consisting of SFPQ, NONO and MATR3.The protein contains 1 matrin type zinc finger and 2 RRM (RNA recognition motif) domains. Two transcript variants encoding the same protein have been identified for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MATR3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat