Anti-AGPAT1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5023R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:1-acyl-sn-glycerol-3-phosphate acyltransferase alpha
- Antigen symbol:AGPAT1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10554
- Antigen synonyms:G15|1-AGPAT1|LPAATA|LPAAT-alpha
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
AGPAT1 converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AGPAT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat