Order Entry
Sweden
ContactUsLinkComponent
 
Anti-RPA32 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-4182R-A350
 :  Bioss
Anti-RPA32 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-4182R-A350
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Replication Protein A32
  • Antigensymbol:
    RPA32
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 350
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    6118
  • Antigen synonymer:
    RPA32|REPA2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    110-160/270
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

As part of the heterotrimeric replication protein A complex (RPA/RP-A), binds and stabilizes single-stranded DNA intermediates, that form during DNA replication or upon DNA stress. It prevents their reannealing and in parallel, recruits and activates different proteins and complexes involved in DNA metabolism. Thereby, it plays an essential role both in DNA replication and the cellular response to DNA damage. In the cellular response to DNA damage, the RPA complex controls DNA repair and DNA damage checkpoint activation. Through recruitment of ATRIP activates the ATR kinase a master regulator of the DNA damage response. It is required for the recruitment of the DNA double-strand break repair factors RAD51 and RAD52 to chromatin in response to DNA damage. Also recruits to sites of DNA damage proteins like XPA and XPG that are involved in nucleotide excision repair and is required for this mechanism of DNA repair. Plays also a role in base excision repair (BER) probably through interaction with UNG. Through RFWD3 may activate CHEK1 and play a role in replication checkpoint control. Also recruits SMARCAL1/HARP, which is involved in replication fork restart, to sites of DNA damage. May also play a role in telomere maintenance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPA32
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat