Order Entry
Sweden
ContactUsLinkComponent
 
Anti-RUNX1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-3023R-CY3
 :  Bioss
Anti-RUNX1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-3023R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Runt-related Transcription Factor 1
  • Antigen symbol:
    RUNX1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    CBFA2|SL3-3 enhancer factor 1 alpha B subunit|PEA2-alpha B|SL3/AKV core-binding factor alpha B subunit|PEBP2-alpha B|RUNX1|Core-binding factor subunit alpha-2|Polyomavirus enhancer-binding protein 2 alpha B subunit|AML1|Oncogene AML-1|CBF-alpha-2|Acute myeloid leukemia 1 protein
  • Modification:
    phospho-Ser249
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

AML1/Runx1 binds DNA as a monomer and through the Runt domain. DNA binding is increased by heterodimerization with CBFB. Isoform AML1L can neither bind DNA nor heterodimerize and interferes with the transactivation activity of AML1/Runx1. CBF binds to the core site, 5'-PYGPYGGT-3', of a number of enhancers and promoters, including murine leukemia virus, polyomavirus enhancer, T cell receptor enhancers, LCK, IL3 and GMCSF promoters. The alpha subunit binds DNA and appears to have a role in the development of normal hematopoiesis. AML1/Runx1 is expressed in a wide variety of tissues and is expressed at the highest levels in thymus, bone marrow and peripheral blood. Defects in AML1/Runx1 are the cause of familial platelet disorder with associated myeloid malignancy, an autosomal dominant disease characterized by qualitative and quantitative platelet defects, and propensity to develop acute myelogenous leukemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RUNX1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat