Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-3997R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 Member A10
- Antigen symbol:SLC25A10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1468
- Antigen synonyms:Solute carrier family 25 member 10|Mitochondrial dicarboxylate carrier|SLC25A10|DIC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:200-250/287
- Purification:Purified by Protein A
- Pk:100 µl
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat