Anti-GRN Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-0823R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Granulin
- Antigen symbol:GRN
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2896
- Antigen synonyms:proepithelin|PGRN|PC cell-derived growth factor|GEP|PCDGF|progranulin|granulin|PEPI|GRN|GP88|acrogranin|CLN11|granulin-epithelin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:442-496/593
- Purification:Purified by Protein A
- Pk:100 µl
Granulins have possible cytokine-like activity. They may play a role in inflammation, wound repair, and tissue remodeling. Granulin-4 promotes proliferation of the epithelial cell line A431 in culture while granulin-3 acts as an antagonist to granulin-4, inhibiting the growth.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Granulin
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat