Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 11B
- Antigen symbol:CDK11B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:984
- Antigen synonyms:p58CDC2L1|CDK11-p46|p58CLK-1|CLK-1|p58|PK58|CDK11|CDC2L1|CDK11-p58|PITSLREA|CDK11-p110|CD2L1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Summary: This gene encodes a member of the p34Cdc2 protein kinase family. p34Cdc2 kinase family members are known to be essential for eukaryotic cell cycle control. This gene is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. The gene loci including this gene, CDC2L2, as well as metalloprotease MMP21/22, consist of two identical, tandemly linked genomic regions which are thought to be a part of the larger region that has been duplicated. This gene and CDC2L2 were shown to be deleted or altered frequently in neuroblastoma with amplified MYCN genes. The protein kinase encoded by this gene could be cleaved by caspases and was demonstrated to play roles in cell apoptosis. Several alternatively spliced variants of this gene have been reported. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDK11B
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: