Anti-CTDSPL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-3509R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CTD (carboxy-terminal Domain, RNA polymerase II, polypeptide A) small phosphatase-like
- Antigen symbol:CTDSPL
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:10217
- Antigen synonyms:PSR1|RBSP3|C3orf8|HYA22|SCP3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-200/276
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Recruited by REST to neuronal genes that contain RE-1 elements, leading to neuronal gene silencing in non-neuronal cells (By similarity). Preferentially catalyzes the dephosphorylation of 'Ser-5' within the tandem 7 residue repeats in the C-terminal domain (CTD) of the largest RNA polymerase II subunit POLR2A. Negatively regulates RNA polymerase II transcription, possibly by controlling the transition from initiation/capping to processive transcript elongation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CTDSPL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: