Anti-KLK11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # BOSSBS-3799R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 11
- Antigen symbol:KLK11
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:11012
- Antigen synonyms:Kallikrein-11 inactive chain 2|Serine protease 20|KLK11|Kallikrein-11 inactive chain 1|PRSS20|Kallikrein11|Hippostasin|UNQ649/PRO1279|Kallikrein 11|TLSP|hK11|Trypsin-like protease|Kallikrein-11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Kallikrein 11 has two alternative splicing isoforms, known as the brain type and prostate type. Elevated serum levels of hK11 have been found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of this biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat