Anti-PTGES3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4020R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin E Synthase 3
- Antigen symbol:PTGES3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10728
- Antigen synonyms:Co chaperone p23|Progesterone receptor complex|Sid 3177|Telomerase binding protein p23|TEBP|Hsp90 co chaperone
- Modification:phospho-Ser113
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Belongs to the p23/wos2 family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat