Anti-NPM Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3309R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nucleophosmin
- Antigen symbol:NPM
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4869
- Antigen synonyms:MGC104254|NO38|Nucleophosmin|Nucleolar protein NO38|numatrin|B23|Nucleolar phosphoprotein B23|NPM1
- Modification:phospho-Thr95
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a phosphoprotein which moves between the nucleus and the cytoplasm. The gene product is thought to be involved in several processes including regulation of the ARF/p53 pathway. A number of genes are fusion partners have been characterized, in particular the anaplastic lymphoma kinase gene on chromosome 2. Mutations in this gene are associated with acute myeloid leukemia. More than a dozen pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.[provided by RefSeq, Nov 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NPM
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat