Anti-CXCR1 Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-1009R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) receptor 1
- Antigen symbol:CXCR1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3577
- Antigen synonyms:CKR-1|C-C-CKR-1|CD181|IL8RBA|IL8RA|CD128|CDw128a|IL8R1|CMKAR1|C-C
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:208-242/350
- Purification:Purified by Protein A
- Pk:100 µl
Receptor to interleukin-8, which is a powerful neutrophils chemotactic factor. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to MGSA (GRO) with a low affinity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CXCR1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: