Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-3542R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:PTEN-induced putative kinase 1 And Polynucleotide Kinase 3'-phosphatase
- Antigen symbol:PNK1/PNKP
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:11284
- Antigen synonyms:PARK6|PNK|PTEN induced putative kinase 1|MCSZ|AOA4|EIEE10|BRPK
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Mammalian PNK catalyzes the phosphorylation of DNA at 5'-hydroxyl termini and can dephosphorylate its 3'-phosphate termini. It plays an important function in DNA repair following ionizing radiation or oxidative damage.PNK has been reported to participate in the repair of DNA-double strand breaks via PARP-1-dependent nonhomologous end-joining.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PNK1/PNKP
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat