Anti-PMP2 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-2032R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peripheral Myelin Protein 2
- Antigen symbol:PMP2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5375
- Antigen synonyms:FABP8|P2|PMP2|Peripheral myelin protein 2|Myelin P2 protein|M-FABP|MP2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
PMP2 is a small basic protein, belonging to the fatty-acid binding protein (FABP) family and is one of the major proteins of peripheral myelin. It appears to be involved in the transport of fatty acids or the metabolism of myelin lipids, and has been shown to have lipid-binding activity. PMP2 protein may have an important role in the organization of compact myelin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PMP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat