Order Entry
Sweden
ContactUsLinkComponent
Anti-PCNA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-PCNA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2007R-A350
Supplier:  Bioss
Anti-PCNA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2007R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proliferating cell nuclear antigen
  • Antigen symbol:
    PCNA
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5111
  • Antigen synonyms:
    DNA polymerase delta auxiliary protein|HGCN8729|MGC8367|Proliferating Cell Nuclear Antigen|cyclin|polymerase delta accessory protein
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    220-261/261
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Auxiliary protein of DNA polymerase delta and is involved in the control of eukaryotic DNA replication by increasing the polymerase's processibility during elongation of the leading strand. Induces a robust stimulatory effect on the 3'-5' exonuclease and 3'-phosphodiesterase, but not apurinic-apyrimidinic (AP) endonuclease, APEX2 activities. Has to be loaded onto DNA in order to be able to stimulate APEX2. Plays a key role in DNA damage response (DDR) by being conveniently positioned at the replication fork to coordinate DNA replication with DNA repair and DNA damage tolerance pathways. Acts as a loading platform to recruit DDR proteins that allow completion of DNA replication after DNA damage and promote postreplication repair: Monoubiquitinated PCNA leads to recruitment of translesion (TLS) polymerases, while 'Lys-63'-linked polyubiquitination of PCNA is involved in error-free pathway and employs recombination mechanisms to synthesize across the lesion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat