Order Entry
Sweden
ContactUsLinkComponent
Anti-GPR78 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-GPR78 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1807R-A647
Supplier:  Bioss
Anti-GPR78 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1807R-A647
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    G Protein-coupled Receptor 78
  • Antigensymbol:
    GPR78
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 647
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    27201
  • Antigen synonymer:
    G-protein coupled receptor 78|UNQ5925/PRO19818|GPR78
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    240-290/363
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

Orphan receptor. Displays a significant level of constitutive activity. Its effect is mediated by G(s)-alpha protein that stimulate adenylate cyclase, resulting in an elevation of intracellular cAMP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR78
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human