Order Entry
Sweden
ContactUsLinkComponent
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-2197R-A647
Supplier:  Bioss
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-2197R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    duck hepatitis A virus 1 Genome polyprotein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    880-910/2249
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

duck hepatitis A virus genome polyprotein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus