Specifications
- Antibody type:Primary
- Antigen name:X-ray repair complementing defective repair in Chinese hamster cells 6
- Antigen symbol:XRCC6
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:2547
- Antigen synonyms:CTC box binding factor 75 kDa subunit|CT|70 kDa subunit of Ku antigen|KU70|TLAA|70 kDa subunit|CTC75|CTCBF|ATP-dependent DNA helicase II|5'-dRP lyase Ku70|5'-deoxyribose-5-phosphate lyase Ku70|ATP-dependent DNA helicase 2 subunit 1|ML8|G22P1|ATP-dependent DNA helicase II 70 kDa subunit
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The p70/p80 autoantigen is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of auto to p70 and p80 have been found in some patients with systemic lupus erythematosus. [provided by RefSeq]. Ku70 heterodimerises with Ku80 to form the ATP-dependent DNA helicase II, a single stranded helicase that binds preferentially to fork-like ends of double-stranded DNA in a cell cycle-dependent manner. The heterodimer plays a role in non-homologous end-joining (NHEJ) required for double-strand break repair and V(D)J recombination. It acts as the regulatory subunit of the DNA-dependent protein kinase complex DNA-PK by increasing the affinity of the catalytic subunit (PRKDC) for DNA. The Ku70/80 heterodimer is also required for osteocalcin gene expression.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: XRCC6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat