Order Entry
Sweden
ContactUsLinkComponent
Anti-FMO5 Rabbit Polyclonal Antibody (Cy3®)
Anti-FMO5 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-13187R-CY3
 :  Bioss
Anti-FMO5 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-13187R-CY3
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Flavin Containing Monooxygenase 5
  • Antigensymbol:
    FMO5
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy3®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    2330
  • Antigen synonymer:
    FMO 5|Hepatic flavin-containing monooxygenase 5|FMO5|Dimethylaniline monooxygenase [N-oxide-forming] 5|Dimethylaniline oxidase 5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    250-300/533
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

In contrast with other forms of FMO it does not seem to be a drug-metabolizing enzyme.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMO5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat