- Antibody type:Primary
- Antigen name:Exosome Component 7
- Antigen symbol:EXOSC7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23016
- Antigen synonyms:Rrp42p|p8|RRP42|EAP1|hRrp42p
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The exosome is a multisubunit complex of 3’ to 5’ exoribonucleases. It is involved in a variety of cellular processes and is responsible for degrading unstable mRNAs that contain AU-rich elements in their untranslated 3’ region. EXOSC7 (exosome component 7), also known as p8, EAP1, RRP42 (Ribosomal RNA-processing protein 42), Rrp42p or hRrp42p, is a component of the exosome multienzyme ribonuclease complex. It belongs to the RNase PH family and localizes to the nucleolus. EXOSC7 is one of the six RNase-PH domain subunits of the exosome. Together, these six subunits form a PNPase-like ring. EXOSC7 is required for the processing of the 7S pre-RNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EXOSC7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat