Specifikationer
- Antibody type:Primary
- Antigen name:Filamin C, Gamma
- Antigen symbol:FLNC
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2318
- Antigen synonyms:FLN2|MFM5|ABPA|ABP-280|ABPL|MPD4|ABP280A
- Modification:phospho-Ser2233
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifikationer
Om denna produkt
Filamins are Actin-binding proteins which contain an N-terminal Actin-binding domain, a membrane glycoprotein domain and a C-terminal self-association domain. Filamins help reshape the cytoskeleton by forming flexible cross-links between two Actin filaments, which maintain membrane integrity during force application. Filamins also participate in signal transduction pathways associated with cell motility, adhesion, differentiation and survival, and force transduction. The filamin family is comprised of Filamin 1, Filamin 2 and Filamin 3. Filamin 2, also designated Filamin C, is a skeletal- and cardiac-muscle specific form of Filamin, which binds ©-sarcoglycan and ∂-sarcoglycan, but not å-sarcoglycan or ∫-sarcoglycan. Muscular dystrophy, an inherited group of disorders resulting in progressive weakness of muscles in the body, is associated with irregular subcellular localization of Filamin 2 caused by a deficiency in KY, a protein that interacts with Filamin 2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FLNC 2233
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat