Order Entry
Sweden
ContactUsLinkComponent
Anti-TMC8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-TMC8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Katalog # BOSSBS-13116R-A647
Supplier:  Bioss
Anti-TMC8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Katalog # BOSSBS-13116R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transmembrane Channel-like 8
  • Antigen symbol:
    TMC8
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    147138
  • Antigen synonyms:
    EV2|Epidermodysplasia verruciformis protein 2|EVIN2|Transmembrane channel-like protein 8|EVER2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Transmembrane channel-like protein 8 (TMC8), also known as Epidermodysplasia verruciformis protein 2 (EVER2), is a 726 amino acid member of the TMC family of proteins. Localized to the endoplasmic reticulum membrane, TMC8 is thought to form a transmembrane channel-like protein with eight predicted transmembrane domains and three leucine zipper motifs. Mutations in the genes encoding TMC8 and TMC6, another member of the TMC family, have been shown to cause epidermodysplasia verruciformis (EV), an autosomal recessive dermatosis characterized by abnormal susceptibility to human papillomaviruses (HPVs) and a high rate of progression to squamous cell carcinoma on sun-exposed skin. Infection by HPVs lead to persistent wart-like or macular lesions. TMC8 is expressed in placenta, prostate and testis, and three named isoforms exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMC8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat