Order Entry
Sweden
ContactUsLinkComponent
Anti-DNM1 Rabbit Polyclonal Antibody (Cy7®)
Anti-DNM1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-13038R-CY7
Supplier:  Bioss
Anti-DNM1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-13038R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dynamin 1
  • Antigen symbol:
    DNM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1759
  • Antigen synonyms:
    EC 3.6.5.5|DNM1|Dynamin BREDNM19|DNM|brain|dynamin-1|Dynamin|B-dynamin|D100
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Dynamin I is a GTPase enzyme required for the retrieval of synaptic vesicles after exocytosis and functions in endocytosis by stimulating assembly of invaginating synaptic vesicles (1). Dynamin I is phosphorylated in nerve terminals exclusively in the cytosolic compartment and in vitro by protein kinase C (PKC) (2–5). The phosphorylation site in PKC-phosphorylated Dynamin I is a single site at Serine 795, which is located near a binding site for the SH3 domain of p85, the regulatory subunit of phosphatidylinositol 3-kinase (2–5). Dephosphorylation is required for synaptic vesicle retrieval, suggesting that phosphorylation affects the subcellular localization of Dynamin I (5). Mouse, rat and human Dynamin I are phosphorylated on serine residues, including Ser 778, by Cdk5, regulating PACSIN1 recruitment and enabling synaptic vesicle endocytosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat