Order Entry
Sweden
ContactUsLinkComponent
Anti-DPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-DPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-13029R-A647
 :  Bioss
Anti-DPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-13029R-A647
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Dipeptidyl-peptidase 3
  • Antigensymbol:
    DPP3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 647
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    10072
  • Antigen synonymer:
    DPPIII
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

Dipeptidyl peptidases (DPPs) mediate regulatory activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. DPPs have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. DPPs can bind specific voltage-gated potassium channels and alter their expression and biophysical properties and may also influence T cells. DPP proteins include DPRP1, DPRP2, DPP3, DPP7, DPP10, DPPX and CD26. DPP3 (dipeptidyl-peptidase 3), also known as DPPIII, is a zinc-exopeptidase that belongs to the peptidase M49 family. DPP3 localizes to the cytoplasm and is involved in intracellular protein catabolism. More specifically, DPP3 is an important enzyme involved in the degradation of enkephalins. An increase in the activity of DPP3 is implicated in ovarian and endometrial cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DPP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat