- Antibody type:Primary
- Antigen name:basic charge, Y-linked, 2B
- Antigen symbol:BPY2B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:442867
- Antigen synonyms:VCY2B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Mammalian spermatogenesis is a complex developmental process. Mutations at multiple loci and in structurally and functionally disparate genes in the genome affect gametogenesis. The analysis of mutations has provided insight into biochemical pathways required for completion of this process. The basic protein on Y chromosome 2 gene (BPY2, also designated VCY2) is located in a frequently deleted azoospermia factor c region. Three copies (paralogs) of the BPY2 gene (BPY2A, BPY2B, BPY2C) reside in the AZFc region. BPY2 protein expression is localized to the nuclei of spermatogonia, spermatocytes and round spermatids, but is absent from elongated spermatids. Impaired expression of BPY2 in infertile men suggests its involvement in male germ cell development. BPY2 interacts with MAP-1S, which shares homology with microtubule-associated proteins (MAPs), suggesting a role for BPY2 within the cytoskeletal network.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BPY2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human