Specifications
- Antibody type:Primary
- Antigen name:Coatomer protein complex, subunit alpha
- Antigen symbol:COPA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1314
- Antigen synonyms:HEPCOP|COPA|Alpha-coat protein|Coatomer subunit alpha|HEP-COP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
COPA (alpha-coat protein) is processed to produce Xenin. Xenin stimulates exocrine pancreatic secretion to affect small and large intestinal motility, and inhibits pentagastrin-stimulated secretion of acid. In the gut, Xenin interacts with the neurotensin receptor. Membrane and vesicular trafficking in the early secretory pathway are mediated by non-Clathrin COP (coat protein) I-coated vesicles. COPI-coated vesicles mediate retrograde transport from the Golgi back to the ER and intra-Golgi transport. The cytosolic precursor of the COPI coat, the heptameric coatomer complex, is composed of two subcomplexes. The first consists of the COPB, COPG, COPD and COPZ subunits (also known as b-, g-, d- and z-COP, respectively), which are distantly homologous to AP Clathrin adaptor subunits. The second consists of the COPA, b'-COP and COPE subunits (also known as a-COP, COPP and e-COP, respectively).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COPA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity: