Anti-ATM Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-12545R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ataxia telangiectasia mutated
- Antigen symbol:ATM
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:472
- Antigen synonyms:ATE|AT1|ATDC|ATD|TEL1|ATC|ATA|TELO1
- Modification:phospho-Ser794
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ATM is a 370 kDa nuclear phosphoprotein involved in the autosomal recessive disease Ataxia Telangiectasia (AT). ATM belongs to a novel family of proteins associated with cell cycle regulation, apoptosis, and response to DNA damage repair (DNA damage caused by such things as ionizing irradiation activates ATM kinase). The C terminal region has extensive homology to the catalytic domains of Phosphatidylinositol 3 kinases (PI3 kinases).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human