Anti-XRCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-1218R-A647
: Bioss
- Antibody type:Primary
- Antigen name:X-ray Repair Complementing Defective Repair in Chinese cells 1
- Antigen symbol:XRCC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7515
- Antigen synonyms:complementing defective|X-ray-repair|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|repair in Chinese hamster
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:13-36/633
- Purification:Purified by Protein A
- Pk:100 µl
Corrects defective DNA strand-break repair and sister chromatid exchange following treatment with ionizing radiation and alkylating agents.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: XRCC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat