Anti-RFX4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-11943R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Regulatory Factor X, 4 (Influences HLA Class II Expression)
- Antigen symbol:RFX4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5992
- Antigen synonyms:RFX4|Testis development protein NYD-SP10|Regulatory factor X 4|Transcription factor RFX4|NYD-SP10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:125-175/735
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May activate transcription by interacting directly with the X-box.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RFX4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat