Order Entry
Sweden
ContactUsLinkComponent
Anti-DCDC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-DCDC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-11824R-HRP
Supplier:  Bioss
Anti-DCDC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-11824R-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Doublecortin Domain Containing 2
  • Antigen symbol:
    DCDC2
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    DCDC2A|NPHP19|RU2|DFNB66|RU2S
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The DCDC2 gene encodes the DCDC2 protein (Doublecortin-containing protein 2, RU2, RU2S) which contains two Doublecortin peptide domains similar to those in the Doublecortin gene. DCDC2 is transcribed as a “normal” gene, which results in a sense transcript (RU2S), but when it is transcribed in the opposite direction, a shorter antisense transcript (RU2AS), which is found in tumors, results. The DCDC2 protein demonstrates ubiquitous expression, whereas RU2AS expression is restricted to normal kidney, bladder, liver and testis, and to tumors of various histologic origins. The deduced DCDC2 protein contains 476 amino acids, while the RU2AS protein contains 84 residues. There is a significant association between dyslexia and several SNPs within the DCDC2 gene.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DCDC2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat