Order Entry
Sweden
ContactUsLinkComponent
Anti-CIC Rabbit Polyclonal Antibody (Cy3®)
Anti-CIC Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11721R-CY3
Supplier:  Bioss
Anti-CIC Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11721R-CY3
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    capicua transcriptional repressor
  • Antigensymbol:
    CIC
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy3®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    23152
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    1490-1540/1608
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

Transcriptional repressor which may play a role in development of the central nervous system (CNS).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CIC
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: