Anti-OR5P3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Katalog # BOSSBS-11628R-A488
Leverantör: Bioss
Specifikationer
- Antibody type:Primary
- Antigen name:Olfactory Receptor, Family 5, Subfamily P Member 3
- Antigen symbol:OR5P3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:120066
- Antigen synonyms:CALN|CCN1|PPP2B|CNA1|CALNA1|JCG1|CALNA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:259-271/311
- Purification:Purified by Protein A
- Pk:100 µl
Specifikationer
Om denna produkt
Odorant receptor (Potential). May be involved in taste perception.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OR5P3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human