Order Entry
Sweden
ContactUsLinkComponent
Anti-NAV1 Rabbit Polyclonal Antibody (Cy5®)
Anti-NAV1 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-11467R-CY5
 :  Bioss
Anti-NAV1 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-11467R-CY5
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Neuron Navigator 1
  • Antigensymbol:
    NAV1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy5®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Antigen synonymer:
    Steerin 1|Protein Steerin 1|POMFIL3|Protein Steerin1|Nav1|Steerin1|mNav1|unc-53 homolog 1|NAV1_HUMAN.|FLJ14203|DKFZp781D0314|Pore membrane and/or filament interacting like protein 3|FLJ12560|Nav 1|POMFIL 3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

Neuron navigator 1 is a 1877 amino acid cytoplasmic protein that is involved in neuronal migration. Neuron navigtor 1 is widely expressed at low levels, though highest expression is found in both adult and fetal nervous tissue. Through interaction with tubulin, Neuron navigator 1 associates with a subset of mirotubule plus ends present in the growth cone. Overexpression of Neuron navigator 1 leads to microtubule bundling, whereas a reduction of its levels causes loss of directionality in the migration of pontine cell leading processes. There are seven isoforms of Neuron navigator 1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuron navigator 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat