Order Entry
Sweden
ContactUsLinkComponent
 
Anti-FLOT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11254R-A350
 :  Bioss
Anti-FLOT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11254R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Flotillin 2
  • Antigen symbol:
    FLOT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2319
  • Antigen synonyms:
    ECS-1|membrane component|ECS1|FLOT2|flotillin 2|reggie-1|surface marker 1 (35kD protein identified by monoclonal ECS-1)|chromosome 17|ESA|M17S1|Flotillin 2 (epidermal surface antigen 1)|ESA1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    180-230/428
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

May act as a scaffolding protein within caveolar membranes, functionally participating in formation of caveolae or caveolae-like vesicles. May be involved in epidermal cell adhesion and epidermal structure and function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECS1/Flotillin 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat