- Antibody type:Primary
- Antigen name:RecQ Protein-like 5
- Antigen symbol:RECQL5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:RECQ5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
RecQ helicases are implicated in DNA recombination and repair helicases and are critical in genome maintenance. RecQL5 is one of five known RecQ homologues in humans, and defects in three of these (BLM, WRN and RecQ4)are associated with autosomal recessive disorders that involve genomic instability and a predisposition to cancer. The exact role of ReqL5 is unclear. It has been shown in mouse embryonic stem (ES) cells that mutations in the Recql5 genes result in a significant increase in the frequency of sister chromatid exchange (SCE), whereas deleting Recql5 lead to an even higher frequency of SCE. There are three different RecQL5 isoforms - alpha, beta and gamma, produced by alternative splicing.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RECQL5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat